2-Formylbenzeneboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Formylphenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 2-Formylphenylboronic acid pinacol ester, 2-(2-Formylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
T3161-1G
T3161-5G
TCT3161-1GEA
45
USD
TCT3161-1G
TCT3161-5G
2-Formylbenzeneboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
2-Formylbenzeneboronic acid pinacol ester
Formula:
C₁₃H₁₇BO₃ MW: 232.08 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07363841 CAS Number: 380151-85-9 |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...