2-Ethoxy-5-methylbenzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 2-Ethoxy-5-methylbenzeneboronic acid, 2-Ethoxy-5-methylphenylboronic acid

E1189-1G E1189-5G
TCE1189-1GEA 85 USD
TCE1189-1G TCE1189-5G
2-Ethoxy-5-methylbenzeneboronic acid (contains varying amounts of Anhydride)
2-Ethoxy-5-methylbenzeneboronic acid
Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
MDL Number: MFCD00196832
CAS Number: 123291-97-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR