2-Cyanobenzeneboronic acid-1,3-propanediol ester ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(1,3,2-Dioxaborinan-2-yl)benzonitrile, 2-Cyanobenzeneboronic acid 1,3-Propanediol ester, 2-Cyanophenylboronic acid 1,3-Propanediol ester

C2993-1G C2993-5G
TCC2993-1GEA 47 USD
TCC2993-1G TCC2993-5G
2-Cyanobenzeneboronic acid-1,3-propanediol ester ≥97.0% (by GC)
2-Cyanobenzeneboronic acid-1,3-propanediol ester
Formula: C₁₀H₁₀BNO₂
MW: 187.00 g/mol
MDL Number: MFCD04039012
CAS Number: 172732-52-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 47.0 to 51.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR