2'-Azido-2'-deoxyuridine ≥98.0% (by HPLC)

Supplier: TCI America

A2942-100MG A2942-25MG
TCA2942-100MGEA 513 USD
TCA2942-100MG TCA2942-25MG
2'-Azido-2'-deoxyuridine ≥98.0% (by HPLC)
2'-Azido-2'-deoxyuridine
Formula: C₉H₁₁N₅O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00043045
CAS Number: 26929-65-7
EINECS: 248-113-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 25.00 to 27.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR