2-(Bromomethyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(2-(bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(Bromomethyl)phenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide

T3393-1G T3393-5G
76220-604EA 123 USD
76220-604 76220-606
2-(Bromomethyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-(Bromomethyl)benzeneboronic acid pinacol ester
Formula: C₁₃H₁₈BBrO₂
MDL Number: MFCD01632210
CAS Number: 377780-72-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR