1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
CTC-Q-PHOS, 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, QPhos, 1,2,3,4,5-Pentaphenyl-1'-(di-t-butylphosphino)ferrocene
P2297-100MG
P2297-500MG
TCP2297-100MGEA
240.6
USD
TCP2297-100MG
TCP2297-500MG
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS ≥98.0% (by titrimetric analysis)
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS
Formula:
C₄₈H₄₇FeP MW: 710.706 g/mol |
MDL Number:
MFCD03788940 CAS Number: 312959-24-3 |
Specification Test Results
Appearance | Orange to Brown powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...