(R)-3-Amino-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A2655-1G
A2655-200MG
TCA2655-1GEA
118
USD
TCA2655-1G
TCA2655-200MG
(R)-3-Amino-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)
(R)-3-Amino-3-phenylpropionic acid
Formula:
C₉H₁₁NO₂ |
MDL Number:
MFCD01076237 CAS Number: 13921-90-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +6.0 to +8.0 deg (C=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...