Graphene, nanoplatelets aggregates, sub-micronparticles surface area 300 m²/g
Supplier: TCI America
|
Warning
|
G0500-25G
G0500-5G
TCG0500-25GEA
176
USD
TCG0500-25G
TCG0500-5G
Graphene, nanoplatelets aggregates, sub-micronparticles surface area 300 m²/g
Graphene
MDL Number:
MFCD00144065 CAS Number: 1034343-98-0 |
Specification Test Results
Appearance | Black powder to crystal |
Specific surface | 300.0 to 350.0 m2/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...