Graphene, nanoplatelets, Length 25 µm, Diameter 6-8 nm

Supplier: TCI America
Warning

G0438-25G G0438-5G
TCG0438-25GEA 358.48 USD
TCG0438-25G TCG0438-5G
Graphene, nanoplatelets, Length 25 µm, Diameter 6-8 nm
Graphene
MDL Number: MFCD00144065
CAS Number: 1034343-98-0

Order Now

Specification Test Results

Appearance Black powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR