Tetrakisacetonitrile copper(I) triflate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Tetrakis(acetonitrile)copper trifluoromethanesulphonate, Tetrakis(acetonitrile)copper(I) Trifluoromethanesulphonate
T3905-1G
T3905-5G
TCT3905-1GEA
123
USD
TCT3905-1G
TCT3905-5G
Tetrakisacetonitrile copper(I) triflate ≥98.0% (by titrimetric analysis)
Tetrakisacetonitrile copper(I) triflate
Formula:
Cu(NCCH₃)₄·CF₃SO₃ MW: 376.821 g/mol Melting Pt: 124°C |
MDL Number:
MFCD10566992 CAS Number: 58452-28-1 |
Specification Test Results
Appearance | White to Light yellow to Green powder to crystal |
Purity (Potassium permanganate method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...