Potassium bis(fluorosulfonyl)imide ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Potassium bis(fluorosulphonyl)amide

P2509-25G P2509-5G
TCP2509-25GEA 219 USD
TCP2509-25G TCP2509-5G
Potassium bis(fluorosulfonyl)imide ≥95.0% (by titrimetric analysis)
Potassium bis(fluorosulfonyl)imide
Formula: F₂KNO₄S₂
MDL Number: MFCD17018950
CAS Number: 14984-76-0
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR