cis-Octahydroisoindole ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
cis-Octahydro-1H-isoindole
O0471-1G
O0471-5G
TCO0471-1GEA
111
USD
TCO0471-1G
TCO0471-5G
cis-Octahydroisoindole ≥97.0% (by GC, titration analysis)
cis-Octahydroisoindole
Formula:
C₈H₁₅N Boiling Pt: 189°C Storage Temperature: Refrigerator |
MDL Number:
MFCD07778620 CAS Number: 1470-99-1 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...