4,4'-Bipyridine -1,1'-dioxide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B6159-1G B6159-250MG
TCB6159-1GEA 163 USD
TCB6159-1G TCB6159-250MG
4,4'-Bipyridine -1,1'-dioxide ≥98.0% (by HPLC, titration analysis)
4,4'-Bipyridine -1,1'-dioxide
Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00134383
CAS Number: 24573-15-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR