2-[(Prop-2-ynyloxy)methyl]oxirane ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2,3-Epoxypropyl Propargyl ether, Glycidyl Propargyl ether, Glycidyl 2-Propynyl ether

G0445-1G G0445-5G
TCG0445-1GEA 30.52 USD
TCG0445-1G TCG0445-5G
2-[(Prop-2-ynyloxy)methyl]oxirane ≥95.0% (by GC)
2-[(Prop-2-ynyloxy)methyl]oxirane
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 95°C
Flash Pt: 69°C
MDL Number: MFCD00099300
CAS Number: 18180-30-8

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR