(3-Bromopropoxy)-tert-butyldimethylsilane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (3-Bromopropoxy)(tert-butyl)dimethylsilane

B5346-25G B5346-5G
76219-578EA 293 USD
76219-578 76219-580
(3-Bromopropoxy)-tert-butyldimethylsilane ≥98.0% (by GC)
(3-Bromopropoxy)-tert-butyldimethylsilane
Formula: C₉H₂₁BrOSi
Boiling Pt: 90°C
Flash Pt: 85°C
Storage Temperature: Refrigerator
MDL Number: MFCD00216589
CAS Number: 89031-84-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR