Benzyl-4-hydroxypiperidine-1-carboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Benzyloxycarbonyl-4-hydroxypiperidine, 4-Hydroxy-1-piperidinecarboxylic acid Benzyl ester, 1-Cbz-4-hydroxypiperidine, 1-Carbobenzoxy-4-piperidinol, 1-Carbobenzoxy-4-hydroxypiperidine, Benzyl 4-Hydroxy-1-piperidinecarboxylate, 1-Benzyloxycarbonyl-4-piperidinol, 1-Cbz-4-piperidinol

B4869-25G B4869-5G
TCB4869-25GEA 162 USD
TCB4869-25G TCB4869-5G
Benzyl-4-hydroxypiperidine-1-carboxylate ≥98.0% (by GC)
Benzyl-4-hydroxypiperidine-1-carboxylate
Formula: C₁₃H₁₇NO₃
MW: 235.29 g/mol
Boiling Pt: 167°C
MDL Number: MFCD01863722
CAS Number: 95798-23-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR