Thiophen-2-carbothioamide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Thiophene-2-carbothioamide

T3604-25G T3604-5G
TCT3604-25GEA 1287 USD
TCT3604-25G TCT3604-5G
Thiophen-2-carbothioamide ≥98.0% (by HPLC)
Thiophen-2-carbothioamide
Formula: C₅H₅NS₂
MW: 143.23 g/mol
MDL Number: MFCD00052890
CAS Number: 20300-02-1

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 101.0 to 105.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR