1-Bromo-2-(methylsulfinyl)benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromo-2-(methylsulphinyl)benzene, 2-Bromophenyl methyll sulphoxide

B4990-1G B4990-5G
TCB4990-1GEA 127 USD
TCB4990-1G TCB4990-5G
1-Bromo-2-(methylsulfinyl)benzene ≥98.0% (by GC)
1-Bromo-2-(methylsulfinyl)benzene
Formula: C₇H₇BrOS
MW: 219.11 g/mol
Boiling Pt: 109°C
MDL Number: MFCD18089235
CAS Number: 7321-58-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR