N-Methyl-N-[4-(trifluoromethyl)phenyl]amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: N-Methyl-4-(trifluoromethyl)aniline

M2774-1G M2774-5G
TCM2774-1GEA 103 USD
TCM2774-1G TCM2774-5G
N-Methyl-N-[4-(trifluoromethyl)phenyl]amine ≥98.0% (by GC, titration analysis)
N-Methyl-N-[4-(trifluoromethyl)phenyl]amine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 106°C
Flash Pt: 98°C
MDL Number: MFCD07369717
CAS Number: 22864-65-9

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR