Tetrachlorophthalonitrile ≥96.0% (by GC, total nitrogen)

Supplier: TCI America
Warning

T3067-25G T3067-5G
TCT3067-25GEA 274.1 USD
TCT3067-25G TCT3067-5G
Tetrachlorophthalonitrile ≥96.0% (by GC, total nitrogen)
Tetrachlorophthalonitrile
Formula: C₈Cl₄N₂
MW: 265.91 g/mol
MDL Number: MFCD00019740
CAS Number: 1953-99-7
EINECS: 217-783-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (with Total Nitrogen) min. 96.0 %
Melting point 249.0 to 253.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR