3-(4-Methylpiperazin-1-yl)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1-(3-Aminophenyl)-4-methylpiperazine, 3-(4-Methyl-1-piperazinyl)aniline

A2714-1G A2714-5G
TCA2714-1GEA 38 USD
TCA2714-1G TCA2714-5G
3-(4-Methylpiperazin-1-yl)aniline ≥98.0% (by GC, titration analysis)
3-(4-Methylpiperazin-1-yl)aniline
Formula: C₁₁H₁₇N₃
MW: 191.28 g/mol
MDL Number: MFCD08435857
CAS Number: 148546-99-0
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR