(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline

P2216-1G P2216-5G
TCP2216-1GEA 17 USD
TCP2216-1G TCP2216-5G
(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline ≥98.0% (by GC, titration analysis)
(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
Formula: C₁₅H₁₅N
MW: 209.29 g/mol
MDL Number: MFCD08692036
CAS Number: 118864-75-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 87.0 to 91.0 deg-C
Specific rotation [a]20/D +10.0 to +14.0 deg (C=0.46, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR