4-(Cyanomethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-(Cyanomethyl)benzeneboronic acid

C3231-1G C3231-5G
TCC3231-1GEA 275 USD
TCC3231-1G TCC3231-5G
4-(Cyanomethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Cyanomethyl)phenylboronic acid
Formula: C₈H₈BNO₂
MW: 160.97 g/mol
Melting Pt: 191°C
MDL Number: MFCD01632200
CAS Number: 91983-26-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 98.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR