2-Difluoromethyl-1H-benzoimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(Difluoromethyl)-1H-benzo[d]imidazole, 2-(Difluoromethyl)benzimidazole

D4763-1G D4763-5G
76219-066EA 44 USD
76219-066 76219-068
2-Difluoromethyl-1H-benzoimidazole ≥98.0% (by GC, titration analysis)
2-Difluoromethyl-1H-benzoimidazole
Formula: C₈H₆F₂N₂
MW: 168.15 g/mol
MDL Number: MFCD00463015
CAS Number: 705-09-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR