4-Oxo-1,4-dihydroquinoline-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1,4-Dihydro-4-oxoquinoline-3-carboxylic acid
D4881-1G
D4881-5G
TCD4881-1GEA
35
USD
TCD4881-1G
TCD4881-5G
4-Oxo-1,4-dihydroquinoline-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
4-Oxo-1,4-dihydroquinoline-3-carboxylic acid
Formula:
C₁₀H₇NO₃ MW: 189.17 g/mol |
MDL Number:
MFCD00498984 CAS Number: 13721-01-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...