4-Methoxy-2,3,6-trimethylbenzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

M3002-1G M3002-5G
TCM3002-1GEA 79 USD
TCM3002-1G TCM3002-5G
4-Methoxy-2,3,6-trimethylbenzaldehyde ≥98.0% (by GC)
4-Methoxy-2,3,6-trimethylbenzaldehyde
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00456729
CAS Number: 54344-92-2
EINECS: 259-117-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR