4-(2-Propynyloxy)benzenecarbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-(Prop-2-yn-1-yloxy)benzaldehyde, 4-(2-Propynyloxy)benzaldehyde, 4-(Propargyloxy)benzaldehyde

P2339-1G P2339-200MG
TCP2339-1GEA 289 USD
TCP2339-1G TCP2339-200MG
4-(2-Propynyloxy)benzenecarbaldehyde ≥98.0% (by GC)
4-(2-Propynyloxy)benzenecarbaldehyde
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD01088150
CAS Number: 5651-86-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR