Chloroformamidine hydrochloride 98,0 N, T
Supplier: TCI America
|
Warning
|
Synonyms:
Carbamimidic chloride hydrochloride
C3199-25G
C3199-5G
TCC3199-25GEA
107
USD
TCC3199-25G
TCC3199-5G
Chloroformamidine hydrochloride 98,0 N, T
Chloroformamidine hydrochloride
Formula:
CH₄Cl₂N₂ MW: 114.96 g/mol Melting Pt: 180°C |
MDL Number:
MFCD00035527 CAS Number: 29671-92-9 EINECS: 249-765-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...