tert-Butyl-3-methyleneazetidine-1-carboxylate ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Methylideneazetidine, N-BOC protected, tert-Butyl 3-methyleneazetidine-1-carboxylate

B6011-1G B6011-5G
TCB6011-1GEA 57 USD
TCB6011-1G TCB6011-5G
tert-Butyl-3-methyleneazetidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-3-methyleneazetidine-1-carboxylate
Formula: C₉H₁₅NO₂
MW: 169.23 g/mol
MDL Number: MFCD12031229
CAS Number: 934664-41-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR