1-Benzhydrylazetidin-3-amine ≥98.0% (by GC)

Supplier: TCI America
Warning

A3283-1G A3283-5G
TCA3283-1GEA 82 USD
TCA3283-1G TCA3283-5G
1-Benzhydrylazetidin-3-amine ≥98.0% (by GC)
1-Benzhydrylazetidin-3-amine
Formula: C₁₆H₁₈N₂
Storage Temperature: Refrigerator
MDL Number: MFCD03093386
CAS Number: 40432-52-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 61.000 to 65.000 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR