3-Propionylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-(Pyridin-3-yl)propan-1-one

P2090-1G P2090-5G
TCP2090-1GEA 42.9 USD
TCP2090-1G TCP2090-5G
3-Propionylpyridine ≥98.0% (by GC)
3-Propionylpyridine
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 77°C
Flash Pt: 103°C
MDL Number: MFCD00023589
CAS Number: 1570-48-5
EINECS: 216-380-8

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR