Oxetan-3-yl 4-methylbenzenesulfonate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
p-Toluenesulphonic acid 3-Oxetanyl ester, 3-Oxetanyl p-Toluenesulphonate, 3-Oxetanyl Tosylate
O0473-25G
O0473-5G
TCO0473-25GEA
323
USD
TCO0473-25G
TCO0473-5G
Oxetan-3-yl 4-methylbenzenesulfonate ≥98.0% (by GC)
Oxetan-3-yl 4-methylbenzenesulfonate
Formula:
C₁₀H₁₂O₄S MW: 228.27 g/mol |
MDL Number:
MFCD08544401 CAS Number: 26272-83-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 89.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...