Tetrahydro-2H-thiopyran-4-ol ≥98.0% (by GC)

Supplier: TCI America

T3373-1G T3373-5G
TCT3373-1GEA 88 USD
TCT3373-1G TCT3373-5G
Tetrahydro-2H-thiopyran-4-ol ≥98.0% (by GC)
Tetrahydro-2H-thiopyran-4-ol
Formula: C₅H₁₀OS
MW: 118.2 g/mol
Boiling Pt: 70°C
MDL Number: MFCD02942489
CAS Number: 29683-23-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR