2-Propylpyridine-4-carbothioamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

P2302-25G P2302-5G
TCP2302-25GEA 143.3 USD
TCP2302-25G TCP2302-5G
2-Propylpyridine-4-carbothioamide ≥98.0% (by HPLC, titration analysis)
2-Propylpyridine-4-carbothioamide
Formula: C₉H₁₂N₂S
MDL Number: MFCD00464119
CAS Number: 14222-60-7
EINECS: 238-093-7
Merck Index: 14,07891

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR