N-(5-Iodo-2-pyridinyl)acetamide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Acetamido-5-iodopyridine, 2-Acetylamino-5-iodopyridine

A3050-1G A3050-5G
TCA3050-1GEA 100 USD
TCA3050-1G TCA3050-5G
N-(5-Iodo-2-pyridinyl)acetamide ≥98.0% (by GC)
N-(5-Iodo-2-pyridinyl)acetamide
Formula: C₇H₇IN₂O
MW: 262.05 g/mol
MDL Number: MFCD01315262
CAS Number: 66131-78-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR