(4-Bromophenyl)(cyclopropyl)ketone ≥95.0% (by GC)

Supplier: TCI America

B4076-1G B4076-5G
TCB4076-1GEA 73 USD
TCB4076-1G TCB4076-5G
(4-Bromophenyl)(cyclopropyl)ketone ≥95.0% (by GC)
(4-Bromophenyl)(cyclopropyl)ketone
Formula: C₁₀H₉BrO
Boiling Pt: 204°C
MDL Number: MFCD00019227
CAS Number: 6952-89-2
EINECS: 230-130-5

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow to Light orange powder to lump to clear liquid
Purity (GC) min. 95.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR