2-(5-Bromo-1H-indol-3-yl)-1-ethanol ≥98.0% (by GC)

Supplier: TCI America

B5068-1G B5068-200MG
TCB5068-1GEA 207.1 USD
TCB5068-1G TCB5068-200MG
2-(5-Bromo-1H-indol-3-yl)-1-ethanol ≥98.0% (by GC)
2-(5-Bromo-1H-indol-3-yl)-1-ethanol
Formula: C₁₀H₁₀BrNO
MDL Number: MFCD00130167
CAS Number: 32774-29-1
EINECS: 251-205-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR