Fmoc-L-β-homoleucine ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Fmoc-L-β-HoLeu-OH, (S)-3-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-5-methylhexanoic acid, Fmoc-L-beta-homoleucine, Fmoc-β-HomoLeu-OH, Fmoc-beta-Homoleu-OH, [(9H-Fluoren-9-ylmethoxy)carbonyl]-L-beta-homoleucine

F1268-1G F1268-250MG
TCF1268-1GEA 231 USD
TCF1268-1G TCF1268-250MG
Fmoc-L-β-homoleucine ≥96.0% (by HPLC, titration analysis)
Fmoc-L-beta-homoleucine
Formula: C₂₂H₂₅NO₄
MW: 367.45 g/mol
Melting Pt: 110°C
Storage Temperature: Refrigerator
MDL Number: MFCD01863059
CAS Number: 193887-44-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Specific rotation [a]20/D -26.0 to -31.0 deg (C=0.15, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR