2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(Prop-2-ynyl)isoindoline-1,3-dione, N-Propargylphthalimide, N-(2-Propynyl)phthalimide

P2329-25G P2329-5G
TCP2329-25GEA 534 USD
TCP2329-25G TCP2329-5G
2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione ≥98.0% (by GC)
2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione
Formula: C₁₁H₇NO₂
MW: 185.18 g/mol
MDL Number: MFCD00065028
CAS Number: 7223-50-9
EINECS: 230-621-4

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR