2-Phenyl-1H-indole-3-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Formyl-2-phenylindole, 2-Phenylindole-3-carboxaldehyde

P2273-1G P2273-5G
TCP2273-1GEA 27 USD
TCP2273-1G TCP2273-5G
2-Phenyl-1H-indole-3-carbaldehyde ≥98.0% (by GC)
2-Phenyl-1H-indole-3-carbaldehyde
Formula: C₁₅H₁₁NO
MW: 221.26 g/mol
MDL Number: MFCD00435481
CAS Number: 25365-71-3

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 252.0 to 256.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR