(3-Chloro-4-methoxyphenyl)acetic acid ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Chloro-4-methoxyphenylacetic acid, 2-(3-Chloro-4-methoxyphenyl)acetic acid
C3311-1G
C3311-5G
TCC3311-1GEA
79
USD
TCC3311-1G
TCC3311-5G
(3-Chloro-4-methoxyphenyl)acetic acid ≥97.0% (by GC)
(3-Chloro-4-methoxyphenyl)acetic acid
Formula:
C₉H₉ClO₃ MW: 200.62 g/mol |
MDL Number:
MFCD02664697 CAS Number: 13721-20-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 92.0 to 97.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...