4-Hydroxy-2-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

H1495-1G H1495-5G
TCH1495-1GEA 151 USD
TCH1495-1G TCH1495-5G
4-Hydroxy-2-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Hydroxy-2-methylphenylboronic acid
Formula: C₇H₉BO₃
MDL Number: MFCD03788424
CAS Number: 493035-82-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR