2-(2-Isopropyl-5-methylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I1014-1G I1014-5G
76220-176EA 29 USD
76220-176 76220-178
2-(2-Isopropyl-5-methylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
2-(2-Isopropyl-5-methylphenoxy)acetic acid
Formula: C₁₂H₁₆O₃
MDL Number: MFCD00021757
CAS Number: 5333-40-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR