2,2'-Biimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

B2487-1G B2487-5G
TCB2487-1GEA 147.5 USD
TCB2487-1G TCB2487-5G
2,2'-Biimidazole ≥98.0% (by HPLC, titration analysis)
2,2'-Biimidazole
Formula: C₆H₆N₄
Melting Pt: 300°C
MDL Number: MFCD00047014
CAS Number: 492-98-8
EINECS: 207-768-8

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR