7-Octyn-1-ol ≥98.0% (by GC)
Supplier: TCI America
O0445-1ML
O0445-5ML
TCO0445-1MLEA
93
USD
TCO0445-1ML
TCO0445-5ML
7-Octyn-1-ol ≥98.0% (by GC)
7-Octyn-1-ol
Formula:
C₈H₁₄O Boiling Pt: 70°C |
MDL Number:
MFCD01632137 CAS Number: 871-91-0 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...