tert-Butyl triphenylphosphoranylidenecarbamate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B5687-25G
B5687-5G
TCB5687-25GEA
419
USD
TCB5687-25G
TCB5687-5G
tert-Butyl triphenylphosphoranylidenecarbamate ≥98.0% (by HPLC, total nitrogen)
tert-Butyl triphenylphosphoranylidenecarbamate
Formula:
C₂₃H₂₄NO₂P |
MDL Number:
MFCD00798174 CAS Number: 68014-21-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...