(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (S)-(+)-Rolipram, (S)-4-(3-(Cyclopentyloxy)-4-methoxyphenyl)pyrrolidin-2-one, (S)-(+)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one

R0183-10MG R0183-50MG
76220-468EA 170 USD
76220-468 76220-470
(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one ≥98.0% (by HPLC)
(4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-pyrrolidin-2-one
Formula: C₁₆H₂₁NO₃
MW: 275.35 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03093861
CAS Number: 85416-73-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 97.0 ee%
Elemental analysis (Nitrogen) 4.50 to 5.50 %
Melting point 132.0 to 136.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR