Etamsylate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,5-Dihydroxybenzenesulphonic acid-N-ethylethanamine, Diethylamine 2,5-Dihydroxybenzenesulphonate

E1145-1G E1145-5G
TCE1145-1GEA 31 USD
TCE1145-1G TCE1145-5G
Etamsylate ≥98.0% (by HPLC, titration analysis)
Etamsylate
Formula: C₁₀H₁₇NO₅S
MW: 263.31 g/mol
MDL Number: MFCD00867499
CAS Number: 2624-44-4
EINECS: 220-090-7
Merck Index: 14,03722

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR