2-[2-[(tert-Butoxycarbonyl)amino]thiazol-4-yl]acetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-BOC-2-amino-4-thiazolacetic acid, 2-(2-((tert-Butoxycarbonyl)amino)thiazol-4-yl)acetic acid, [2-[[[(2-Methyl-2-propanyl)oxy]carbonyl]amino]-1,3-thiazol-4-yl]acetic acid
B6173-1G
B6173-250MG
TCB6173-1GEA
116
USD
TCB6173-1G
TCB6173-250MG
2-[2-[(tert-Butoxycarbonyl)amino]thiazol-4-yl]acetic acid ≥98.0% (by HPLC, titration analysis)
2-[2-[(tert-Butoxycarbonyl)amino]thiazol-4-yl]acetic acid
Formula:
C₁₀H₁₄N₂O₄S |
MDL Number:
MFCD02682394 CAS Number: 89336-46-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 166.0 to 170.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...