Fmoc-L-4-pyridylalanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Fmoc-3-(4-pyridyl)-L-alanine, Fmoc-3-(4-pyridyl)-Ala-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine, Fmoc-4-Pal-OH

F0927-1G F0927-5G
TCF0927-1GEA 146 USD
TCF0927-1G TCF0927-5G
Fmoc-L-4-pyridylalanine ≥98.0% (by HPLC, titration analysis)
Fmoc-L-4-pyridylalanine
Formula: C₂₃H₂₀N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00672566
CAS Number: 169555-95-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR